The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079821.2 | NP_001073289.1 | 5 - 6 | 2291 | 60 |
NM_001127461.2 | NP_001120933.1 | 4 - 5 | 2933 | 60 | |
NM_001243133.1 | NP_001230062.1 | 3 - 4 | 2899 | 60 | |
NM_004895.4 | NP_004886.3 | 3 - 4 | 2899 | 60 | |
XM_011544048.2 | XP_011542350.1 | 4 - 5 | 3038 | 60 | |
XM_011544053.2 | XP_011542355.1 | 4 - 5 | 2926 | 60 | |
XM_017000181.1 | XP_016855670.1 | 5 - 6 | 2515 | 60 | |
XM_017000182.1 | XP_016855671.1 | 5 - 6 | 2460 | 60 | |
GenBank mRNA | AB208891.1 | - | 3 - 4 | 2908 | 60 |
AF410477.1 | - | 3 - 4 | 2899 | 60 | |
AF420469.1 | - | 4 - 5 | 2285 | 60 | |
AF468522.1 | - | 3 - 4 | 2315 | 60 | |
AK314998.1 | - | 5 - 6 | 2291 | 60 | |
BC143359.1 | - | 3 - 4 | 2218 | 60 | |
BC143362.1 | - | 3 - 4 | 2218 | 60 |