Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAV2
Entrez Gene ID
89797
Gene Name
neuron navigator 2
Gene Aliases
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location
Chr.11: 19345200 - 20121601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111018.1 NP_001104488.1 12 - 13 2944 65
NM_001111019.2 NP_001104489.1 3 - 4 422 65
NM_001244963.1 NP_001231892.1 13 - 14 3337 65
NM_145117.4 NP_660093.2 12 - 13 3268 65
NM_182964.5 NP_892009.3 12 - 13 3268 65
XM_005253214.4 XP_005253271.1 2 - 3 4854 65
XM_006718364.3 XP_006718427.1 12 - 13 3590 65
XM_006718365.3 XP_006718428.1 13 - 14 3659 65
XM_006718366.3 XP_006718429.1 13 - 14 2823 65
XM_006718367.3 XP_006718430.1 2 - 3 4857 65
XM_006718368.3 XP_006718431.1 2 - 3 4854 65
XM_006718369.2 XP_006718432.1 2 - 3 352 65
XM_011520444.2 XP_011518746.1 13 - 14 3659 65
XM_011520445.2 XP_011518747.1 13 - 14 3659 65
XM_011520446.2 XP_011518748.1 13 - 14 3659 65
XM_011520447.2 XP_011518749.1 13 - 14 3659 65
XM_011520448.2 XP_011518750.1 13 - 14 3530 65
XM_011520449.2 XP_011518751.1 13 - 14 3659 65
XM_011520450.2 XP_011518752.1 13 - 14 3659 65
XM_011520451.2 XP_011518753.1 12 - 13 3037 65
XM_011520452.2 XP_011518754.1 13 - 14 3407 65
XM_011520453.2 XP_011518755.1 2 - 3 287 65
XM_011520454.2 XP_011518756.1 2 - 3 287 65
XM_017018520.1 XP_016874009.1 14 - 15 3451 65
XM_017018521.1 XP_016874010.1 13 - 14 3659 65
XM_017018522.1 XP_016874011.1 13 - 14 3407 65
XM_017018523.1 XP_016874012.1 13 - 14 4336 65
XM_017018524.1 XP_016874013.1 13 - 14 3159 65
XM_017018525.1 XP_016874014.1 7 - 8 2002 65
XM_017018526.1 XP_016874015.1 3 - 4 2638 65
GenBank mRNA AB037840.3 - 12 - 13 3269 65
AB063115.1 - 12 - 13 2944 65
AB063116.1 - 12 - 13 3237 65
AF466143.1 - 12 - 13 2983 65
AF466144.1 - 12 - 13 2983 65
AJ488102.1 - 13 - 14 3075 65
AK074287.1 - 3 - 4 422 65
AK298346.1 - 2 - 3 434 65
AK307574.1 - 2 - 3 250 65
BC150290.1 - 12 - 13 3269 65

View Full Product Details