Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
Chromosome Location
Chr.5: 129460281 - 129738683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543246.2 XP_011541548.1 7 - 8 851 90
XM_011543247.2 XP_011541549.1 7 - 8 875 90
XM_011543248.2 XP_011541550.1 7 - 8 995 90
XM_011543249.2 XP_011541551.1 2 - 3 1154 90
XM_017009174.1 XP_016864663.1 6 - 7 1610 90
XM_017009175.1 XP_016864664.1 3 - 4 1907 90
XM_017009176.1 XP_016864665.1 8 - 9 1035 90
GenBank mRNA AJ311904.1 - 8 - 9 1463 90
AK128729.1 - 6 - 7 678 90

View Full Product Details