The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138635.3 | NP_619541.1 | 4 - 5 | 483 | 102 |
GenBank mRNA | AK023973.1 | - | 4 - 5 | 434 | 102 |
BM452844.1 | - | 4 - 5 | 364 | 102 |