The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013310.3 | NP_037442.3 | 3 - 4 | 432 | 64 |
GenBank mRNA | AF038169.1 | - | 3 - 4 | 432 | 64 |
BC071972.1 | - | 2 - 3 | 226 | 64 | |
BC093716.1 | - | 1 - 2 | 52 | 64 | |
BC112189.1 | - | 1 - 2 | 52 | 64 |