The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130806.3 | NP_570718.1 | 10 - 11 | 927 | 117 |
XM_017020389.1 | XP_016875878.1 | 10 - 11 | 916 | 117 | |
GenBank mRNA | AF403384.2 | - | 10 - 11 | 962 | 117 |
AF453828.1 | - | 10 - 11 | 926 | 117 |