The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166058.1 | NP_001159530.1 | 16 - 17 | 2004 | 119 |
NM_130806.3 | NP_570718.1 | 17 - 18 | 2076 | 119 | |
GenBank mRNA | AF403384.2 | - | 17 - 18 | 2111 | 119 |
AF453828.1 | - | 17 - 18 | 2075 | 119 | |
AY899851.1 | - | 16 - 17 | 1933 | 119 |