Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS16
Entrez Gene ID
170690
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 16
Gene Aliases
ADAMTS16s
Chromosome Location
Chr.5: 5140330 - 5320304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139056.3 NP_620687.2 8 - 9 1448 96
NR_136935.1 8 - 9 1448 96
GenBank mRNA AB095949.1 - 5 - 6 703 96
AJ315734.1 - 8 - 9 1310 96
AK122980.1 - 6 - 7 1073 96
AK127746.1 - 8 - 9 1448 96
DQ266047.1 - 8 - 9 1448 96

View Full Product Details