Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB2B
Entrez Gene ID
84932
Gene Name
RAB2B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.14: 21459020 - 21476973 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032846.3 NP_116235.2 1 - 2 146 83
NR_028074.1 1 - 2 146 83
GenBank mRNA AF468652.1 - 1 - 2 139 83
AK027730.1 - 1 - 2 53 83
AK296057.1 - 1 - 2 56 83
AK307349.1 - 1 - 2 131 83
AK315354.1 - 1 - 2 126 83
BC020839.1 - 1 - 2 135 83
BC066366.1 - 1 - 2 113 83
BX647601.1 - 1 - 2 107 83
CD634472.1 - 1 - 2 146 83

View Full Product Details