The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181703.3 | NP_859054.1 | 1 - 2 | 110 | 60 |
XM_017001044.1 | XP_016856533.1 | 2 - 3 | 2152 | 60 | |
GenBank mRNA | BC013313.2 | - | 1 - 2 | 81 | 60 |
BX377692.2 | - | 1 - 2 | 110 | 60 |