The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271999.1 | NP_001258928.1 | 38 - 39 | 5006 | 105 |
NM_001272000.1 | NP_001258929.1 | 37 - 38 | 4913 | 105 | |
NM_001272001.1 | NP_001258930.1 | 37 - 38 | 4913 | 105 | |
NM_033407.3 | NP_212132.2 | 37 - 38 | 4940 | 105 | |
XM_005271292.2 | XP_005271349.1 | 38 - 39 | 4996 | 105 | |
XM_011542326.2 | XP_011540628.1 | 38 - 39 | 5023 | 105 | |
XM_011542327.2 | XP_011540629.1 | 38 - 39 | 5023 | 105 | |
XM_011542328.2 | XP_011540630.1 | 38 - 39 | 5023 | 105 | |
XM_011542330.2 | XP_011540632.1 | 38 - 39 | 5023 | 105 | |
XM_017002639.1 | XP_016858128.1 | 37 - 38 | 4930 | 105 | |
XM_017002640.1 | XP_016858129.1 | 38 - 39 | 5023 | 105 | |
GenBank mRNA | AB051558.1 | - | 18 - 19 | 2416 | 105 |
AK055493.1 | - | 4 - 5 | 557 | 105 | |
AK055905.1 | - | 10 - 11 | 1439 | 105 | |
AK091479.1 | - | 11 - 12 | 1407 | 105 | |
AK125049.1 | - | 23 - 24 | 3085 | 105 | |
AK299737.1 | - | 9 - 10 | 1123 | 105 | |
BC016392.1 | - | 7 - 8 | 910 | 105 | |
DQ118679.1 | - | 34 - 35 | 4485 | 105 | |
DQ118680.1 | - | 34 - 35 | 4485 | 105 | |
DQ341187.1 | - | 38 - 39 | 4913 | 105 |