Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAPVD1
Entrez Gene ID
26130
Gene Name
GTPase activating protein and VPS9 domains 1
Gene Aliases
GAPEX5, GAPex-5, RAP6
Chromosome Location
Chr.9: 125261794 - 125365011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282679.1 NP_001269608.1 8 - 9 1810 94
NM_001282680.1 NP_001269609.1 8 - 9 1810 94
NM_001282681.1 NP_001269610.1 5 - 6 1478 94
NM_015635.3 NP_056450.2 6 - 7 1644 94
XM_005251901.3 XP_005251958.1 7 - 8 1674 94
XM_005251904.3 XP_005251961.1 7 - 8 1671 94
XM_006717044.3 XP_006717107.1 7 - 8 1659 94
XM_011518499.2 XP_011516801.1 7 - 8 1688 94
XM_011518500.2 XP_011516802.1 8 - 9 1805 94
XM_011518502.2 XP_011516804.1 7 - 8 1616 94
XM_011518506.2 XP_011516808.1 7 - 8 1676 94
XM_011518507.2 XP_011516809.1 7 - 8 1693 94
XM_017014600.1 XP_016870089.1 6 - 7 1498 94
XM_017014601.1 XP_016870090.1 6 - 7 1601 94
XM_017014602.1 XP_016870091.1 8 - 9 1821 94
XM_017014603.1 XP_016870092.1 8 - 9 1768 94
XM_017014604.1 XP_016870093.1 9 - 10 1870 94
XM_017014605.1 XP_016870094.1 6 - 7 1601 94
XM_017014606.1 XP_016870095.1 8 - 9 1778 94
XM_017014607.1 XP_016870096.1 8 - 9 1787 94
XM_017014608.1 XP_016870097.1 6 - 7 1597 94
XM_017014609.1 XP_016870098.1 8 - 9 1783 94
XM_017014610.1 XP_016870099.1 6 - 7 1601 94
XM_017014611.1 XP_016870100.1 7 - 8 1657 94
GenBank mRNA AB040954.3 - 8 - 9 1736 94
AK000126.1 - 4 - 5 1034 94
AK027049.1 - 4 - 5 496 94
AK056493.1 - 6 - 7 1606 94
AK294542.1 - 6 - 7 1644 94
AK296381.1 - 7 - 8 1734 94
BC010027.2 - 5 - 6 1483 94
BC021119.1 - 4 - 5 1135 94
BC114937.1 - 8 - 9 1693 94
BC114962.1 - 6 - 7 1529 94

View Full Product Details