The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130720.2 | NP_001124192.1 | 7 - 8 | 1085 | 62 |
NM_001130722.2 | NP_001124194.2 | 7 - 8 | 943 | 62 | |
NM_001130723.2 | NP_001124195.1 | 7 - 8 | 1088 | 62 | |
NM_015328.3 | NP_056143.1 | 7 - 8 | 1088 | 62 | |
XM_005250237.1 | XP_005250294.1 | 7 - 8 | 1091 | 62 | |
XM_011515987.2 | XP_011514289.1 | 8 - 9 | 2062 | 62 | |
XM_011515988.2 | XP_011514290.1 | 8 - 9 | 2029 | 62 | |
XM_017011904.1 | XP_016867393.1 | 7 - 8 | 806 | 62 | |
XM_017011905.1 | XP_016867394.1 | 7 - 8 | 966 | 62 | |
XM_017011906.1 | XP_016867395.1 | 8 - 9 | 982 | 62 | |
XM_017011907.1 | XP_016867396.1 | 8 - 9 | 790 | 62 | |
GenBank mRNA | AB020635.1 | - | 7 - 8 | 1049 | 62 |
AK025372.1 | - | 7 - 8 | 987 | 62 | |
AK295851.1 | - | 7 - 8 | 777 | 62 | |
AK316073.1 | - | 7 - 8 | 943 | 62 | |
BC008349.1 | - | 7 - 8 | 1027 | 62 | |
BC024325.1 | - | 7 - 8 | 1079 | 62 |