The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128212.2 | NP_001121684.1 | 1 - 2 | 215 | 111 |
NM_001307994.1 | NP_001294923.1 | 1 - 2 | 215 | 111 | |
XM_005246355.2 | XP_005246412.2 | 1 - 2 | 209 | 111 | |
XM_017003481.1 | XP_016858970.1 | 1 - 2 | 209 | 111 | |
XM_017003482.1 | XP_016858971.1 | 1 - 2 | 209 | 111 | |
XM_017003483.1 | XP_016858972.1 | 1 - 2 | 124 | 111 | |
GenBank mRNA | BC029520.1 | - | 1 - 2 | 119 | 111 |
DR000438.1 | - | 1 - 2 | 222 | 111 |