The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244949.1 | NP_001231878.1 | 9 - 10 | 1005 | 112 |
NM_020918.5 | NP_065969.3 | 9 - 10 | 1001 | 112 | |
XM_005269998.1 | XP_005270055.1 | 9 - 10 | 1146 | 112 | |
GenBank mRNA | AB046780.1 | - | 3 - 4 | 293 | 112 |
AK172799.1 | - | 7 - 8 | 834 | 112 | |
AL832464.1 | - | 9 - 10 | 957 | 112 | |
AL833061.2 | - | 9 - 10 | 987 | 112 | |
AL833093.2 | - | 9 - 10 | 990 | 112 | |
BC030783.1 | - | 9 - 10 | 969 | 112 | |
BX647451.1 | - | 9 - 10 | 969 | 112 |