Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR88
Entrez Gene ID
126248
Gene Name
WD repeat domain 88
Gene Aliases
PQWD
Chromosome Location
Chr.19: 33132092 - 33175799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_135565.1 10 - 11 1317 127
GenBank mRNA BC031227.1 - 10 - 11 1317 127

View Full Product Details