The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020750.2 | NP_065801.1 | 8 - 9 | 1123 | 111 |
GenBank mRNA | AB033117.2 | - | 8 - 9 | 1062 | 111 |
AF271159.1 | - | 8 - 9 | 912 | 111 | |
AF298880.1 | - | 8 - 9 | 975 | 111 | |
AK291452.1 | - | 8 - 9 | 1086 | 111 | |
AL834207.1 | - | 8 - 9 | 1032 | 111 | |
BC009969.2 | - | 1 - 2 | 87 | 111 | |
BC062635.1 | - | 8 - 9 | 1089 | 111 |