The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032228.5 | NP_115604.1 | 3 - 4 | 519 | 68 |
XM_011520400.1 | XP_011518702.1 | 3 - 4 | 519 | 68 | |
GenBank mRNA | AK315527.1 | - | 3 - 4 | 468 | 68 |
AY358784.1 | - | 3 - 4 | 508 | 68 | |
AY423606.1 | - | 3 - 4 | 462 | 68 | |
AY600449.1 | - | 3 - 4 | 454 | 68 | |
BC017377.2 | - | 3 - 4 | 461 | 68 | |
CR936619.1 | - | 3 - 4 | 494 | 68 |