The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015347.4 | NP_056162.4 | 14 - 15 | 2811 | 75 |
XM_011538102.1 | XP_011536404.1 | 16 - 17 | 3629 | 75 | |
XM_011538103.2 | XP_011536405.1 | 18 - 19 | 4091 | 75 | |
XM_011538104.2 | XP_011536406.1 | 18 - 19 | 3906 | 75 | |
XM_011538105.2 | XP_011536407.1 | 17 - 18 | 3363 | 75 | |
XM_011538106.2 | XP_011536408.1 | 16 - 17 | 3204 | 75 | |
XM_017019105.1 | XP_016874594.1 | 18 - 19 | 4104 | 75 | |
XM_017019106.1 | XP_016874595.1 | 17 - 18 | 3363 | 75 | |
XM_017019107.1 | XP_016874596.1 | 17 - 18 | 3185 | 75 | |
XM_017019108.1 | XP_016874597.1 | 17 - 18 | 3368 | 75 | |
XM_017019109.1 | XP_016874598.1 | 15 - 16 | 2912 | 75 | |
XM_017019110.1 | XP_016874599.1 | 17 - 18 | 3185 | 75 | |
XM_017019111.1 | XP_016874600.1 | 17 - 18 | 3185 | 75 | |
GenBank mRNA | AB002316.1 | - | 14 - 15 | 2811 | 75 |
AY699979.1 | - | 11 - 12 | 2502 | 75 | |
BX641152.1 | - | 1 - 2 | 7543 | 75 |