The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015164.2 | NP_055979.2 | 11 - 12 | 2149 | 69 |
XM_005245790.3 | XP_005245847.1 | 10 - 11 | 2284 | 69 | |
XM_005245791.4 | XP_005245848.1 | 6 - 7 | 1417 | 69 | |
XM_017000757.1 | XP_016856246.1 | 11 - 12 | 2056 | 69 | |
XM_017000758.1 | XP_016856247.1 | 10 - 11 | 1996 | 69 | |
GenBank mRNA | AB020649.1 | - | 11 - 12 | 1925 | 69 |
BC008002.1 | - | 3 - 4 | 186 | 69 | |
BC016488.1 | - | 3 - 4 | 831 | 69 | |
BC030545.1 | - | 2 - 3 | 129 | 69 | |
BC040441.1 | - | 11 - 12 | 2152 | 69 | |
BC042103.1 | - | 3 - 4 | 479 | 69 | |
BC068599.1 | - | 3 - 4 | 504 | 69 |