The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020745.3 | NP_065796.1 | 5 - 6 | 900 | 77 |
XM_011514764.2 | XP_011513066.1 | 5 - 6 | 932 | 77 | |
GenBank mRNA | AB033096.1 | - | 5 - 6 | 900 | 77 |
BC033169.1 | - | 2 - 3 | 297 | 77 | |
BC131728.1 | - | 5 - 6 | 900 | 77 |