The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014431.2 | NP_055246.2 | 8 - 9 | 1304 | 104 |
XM_011539637.2 | XP_011537939.1 | 8 - 9 | 2490 | 104 | |
XM_011539638.2 | XP_011537940.1 | 8 - 9 | 1077 | 104 | |
XM_017016072.1 | XP_016871561.1 | 8 - 9 | 1075 | 104 | |
GenBank mRNA | AB033100.2 | - | 8 - 9 | 1302 | 104 |
AK021806.1 | - | 4 - 5 | 525 | 104 | |
BC040163.1 | - | 6 - 7 | 807 | 104 | |
BC136375.1 | - | 8 - 9 | 1186 | 104 | |
BC136376.1 | - | 8 - 9 | 1186 | 104 |