The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014431.2 | NP_055246.2 | 17 - 18 | 2402 | 74 |
XM_011539637.2 | XP_011537939.1 | 17 - 18 | 3588 | 74 | |
XM_011539638.2 | XP_011537940.1 | 17 - 18 | 2175 | 74 | |
XM_017016072.1 | XP_016871561.1 | 17 - 18 | 2173 | 74 | |
GenBank mRNA | AB033100.2 | - | 17 - 18 | 2400 | 74 |
BC015004.1 | - | 3 - 4 | 262 | 74 | |
BC040163.1 | - | 15 - 16 | 1905 | 74 | |
BC136375.1 | - | 17 - 18 | 2284 | 74 | |
BC136376.1 | - | 17 - 18 | 2284 | 74 |