Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE2
Entrez Gene ID
23224
Gene Name
spectrin repeat containing nuclear envelope protein 2
Gene Aliases
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location
Chr.14: 63761904 - 64226449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015180.4 NP_055995.4 95 - 96 17663 88
NM_182914.2 NP_878918.2 95 - 96 17663 88
XM_005267454.1 XP_005267511.1 95 - 96 17663 88
XM_005267456.1 XP_005267513.1 95 - 96 17663 88
XM_005267457.1 XP_005267514.1 95 - 96 17663 88
XM_005267458.1 XP_005267515.1 95 - 96 17663 88
XM_005267459.1 XP_005267516.1 95 - 96 17663 88
XM_011536574.1 XP_011534876.1 95 - 96 17663 88
XM_011536575.2 XP_011534877.1 95 - 96 17640 88
XM_011536576.2 XP_011534878.1 96 - 97 17820 88
XM_011536577.2 XP_011534879.1 95 - 96 17587 88
XM_011536578.1 XP_011534880.1 95 - 96 17663 88
XM_011536579.1 XP_011534881.1 95 - 96 17663 88
XM_011536580.1 XP_011534882.1 95 - 96 17663 88
XM_011536581.1 XP_011534883.1 95 - 96 17663 88
XM_011536582.1 XP_011534884.1 94 - 95 17546 88
XM_017021101.1 XP_016876590.1 95 - 96 17665 88
XM_017021102.1 XP_016876591.1 94 - 95 17708 88
GenBank mRNA AB023228.3 - 46 - 47 7335 88
AF357236.1 - 93 - 94 17049 88
AF435011.1 - 95 - 96 17645 88
AF495911.1 - 95 - 96 17630 88
AK074055.1 - 8 - 9 1297 88
AK090430.1 - 13 - 14 2039 88
AK095087.1 - 21 - 22 3401 88
AL080133.1 - 1 - 2 176 88
AY061759.1 - 43 - 44 6966 88
BX537642.1 - 16 - 17 2476 88
BX538095.1 - 4 - 5 597 88
CR749324.1 - 15 - 16 2360 88

View Full Product Details