Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RANBP10
Entrez Gene ID
57610
Gene Name
RAN binding protein 10
Gene Aliases
-
Chromosome Location
Chr.16: 67723066 - 67806652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320238.1 NP_001307167.1 4 - 5 594 77
NM_001320240.1 NP_001307169.1 8 - 9 1159 77
NM_020850.2 NP_065901.1 5 - 6 706 77
XM_005256070.2 XP_005256127.1 5 - 6 614 77
XM_005256072.2 XP_005256129.1 4 - 5 561 77
XM_017023508.1 XP_016878997.1 6 - 7 774 77
XM_017023509.1 XP_016878998.1 3 - 4 209 77
GenBank mRNA AB040897.1 - 5 - 6 595 77
AK055727.1 - 8 - 9 1064 77
AK295171.1 - 4 - 5 561 77
AK295530.1 - 4 - 5 502 77
AK304030.1 - 5 - 6 611 77
AY337313.1 - 5 - 6 706 77
BC099917.1 - 5 - 6 611 77
BC121176.1 - 5 - 6 704 77
BC121177.2 - 5 - 6 704 77

View Full Product Details