Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPP7
Entrez Gene ID
143098
Gene Name
membrane palmitoylated protein 7
Gene Aliases
-
Chromosome Location
Chr.10: 28050993 - 28303066 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318170.1 NP_001305099.1 11 - 12 1285 147
NM_173496.4 NP_775767.2 13 - 14 1311 147
NR_134517.1 11 - 12 1285 147
NR_134518.1 10 - 11 1166 147
XM_011519337.2 XP_011517639.1 12 - 13 1210 147
XM_011519338.2 XP_011517640.1 10 - 11 1166 147
XM_017015741.1 XP_016871230.1 10 - 11 1923 147
XM_017015742.1 XP_016871231.1 11 - 12 1224 147
GenBank mRNA AK057360.2 - 11 - 12 1208 147
AK301592.1 - 10 - 11 1166 147
AK315046.1 - 11 - 12 1230 147
AL832380.1 - 10 - 11 1081 147
BC038105.2 - 13 - 14 1303 147

View Full Product Details