Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAMD1A
Entrez Gene ID
57655
Gene Name
GRAM domain containing 1A
Gene Aliases
KIAA1533
Chromosome Location
Chr.19: 34994784 - 35026471 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136199.2 NP_001129671.1 15 - 16 1980 103
NM_001320034.1 NP_001306963.1 16 - 17 2001 103
NM_001320035.1 NP_001306964.1 15 - 16 1888 103
NM_001320036.1 NP_001306965.1 17 - 18 2171 103
XM_017027035.1 XP_016882524.1 16 - 17 2118 103
GenBank mRNA AK026064.1 - 7 - 8 818 103
AK074827.1 - 13 - 14 1564 103
AK074864.1 - 15 - 16 1890 103
AK074914.1 - 15 - 16 1874 103
AK097215.1 - 17 - 18 2171 103
BC041618.1 - 6 - 7 1628 103

View Full Product Details