Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRAM2
Entrez Gene ID
128338
Gene Name
DNA damage regulated autophagy modulator 2
Gene Aliases
CORD21, PRO180, TMEM77, WWFQ154
Chromosome Location
Chr.1: 111117332 - 111140216 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005270469.2 XP_005270526.1 2 - 3 236 121
XM_005270470.1 XP_005270527.1 2 - 3 359 121
XM_006710361.1 XP_006710424.1 2 - 3 308 121
XM_006710362.1 XP_006710425.1 2 - 3 281 121
XM_017000320.1 XP_016855809.1 1 - 2 232 121
GenBank mRNA AL833098.1 - 1 - 2 546 121
BC091509.1 - 2 - 3 270 121

View Full Product Details