Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRK
Entrez Gene ID
5796
Gene Name
protein tyrosine phosphatase, receptor type K
Gene Aliases
R-PTP-kappa
Chromosome Location
Chr.6: 127968779 - 128520847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135648.2 NP_001129120.1 2 - 3 541 81
NM_001291981.1 NP_001278910.1 2 - 3 541 81
NM_001291982.1 NP_001278911.1 2 - 3 541 81
NM_001291984.1 NP_001278913.1 2 - 3 541 81
NM_002844.3 NP_002835.2 2 - 3 541 81
XM_006715537.2 XP_006715600.1 2 - 3 592 81
XM_011536014.2 XP_011534316.1 2 - 3 592 81
XM_011536015.2 XP_011534317.1 2 - 3 592 81
XM_011536016.2 XP_011534318.1 2 - 3 234 81
XM_011536017.2 XP_011534319.1 3 - 4 832 81
XM_011536018.2 XP_011534320.1 3 - 4 505 81
XM_011536020.2 XP_011534322.1 2 - 3 592 81
XM_011536021.2 XP_011534323.1 2 - 3 592 81
XM_017011145.1 XP_016866634.1 3 - 4 556 81
XM_017011146.1 XP_016866635.1 3 - 4 505 81
XM_017011147.1 XP_016866636.1 3 - 4 486 81
XM_017011148.1 XP_016866637.1 3 - 4 483 81
GenBank mRNA AF533875.1 - 2 - 3 306 81
AK295028.1 - 2 - 3 466 81
BC063596.1 - 3 - 4 550 81
BC140775.1 - 2 - 3 343 81
BC144512.1 - 2 - 3 343 81
BC144513.1 - 2 - 3 343 81
BI755683.1 - - 446 81
CR749277.1 - 2 - 3 482 81
L77886.1 - 2 - 3 240 81
Z70660.1 - 2 - 3 225 81

View Full Product Details