The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251825.1 | NP_001238754.1 | 3 - 4 | 1631 | 62 |
NM_003109.1 | NP_003100.1 | 3 - 4 | 1712 | 62 | |
NM_138473.2 | NP_612482.2 | 3 - 4 | 1775 | 62 | |
XM_011538696.2 | XP_011536998.1 | 2 - 3 | 1690 | 62 | |
GenBank mRNA | AB209678.1 | - | 1 - 2 | 1014 | 62 |
AF252284.1 | - | 2 - 3 | 1669 | 62 | |
BC043224.1 | - | 3 - 4 | 1717 | 62 | |
BC062539.1 | - | 3 - 4 | 1776 | 62 | |
FJ949572.1 | - | 1 - 2 | 220 | 62 | |
FN908228.1 | - | 2 - 3 | 1534 | 62 | |
J03133.1 | - | 1 - 2 | 1411 | 62 |