The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032901.3 | NP_116290.1 | 1 - 2 | 322 | 64 |
GenBank mRNA | AK312180.1 | - | 1 - 2 | 105 | 64 |
BC007849.2 | - | 1 - 2 | 186 | 64 | |
BP369305.1 | - | 1 - 2 | 321 | 64 |