The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152755.1 | NP_689968.1 | 1 - 2 | 225 | 83 |
GenBank mRNA | AK075537.1 | - | 1 - 2 | 179 | 83 |
AY359100.1 | - | 1 - 2 | 229 | 83 | |
BC019903.1 | - | 1 - 2 | 155 | 83 | |
BC032339.1 | - | 1 - 2 | 225 | 83 |