The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015344.2 | NP_056159.2 | 3 - 4 | 386 | 106 |
GenBank mRNA | AF063605.1 | - | 3 - 4 | 363 | 106 |
AF161461.1 | - | 3 - 4 | 319 | 105 | |
AK075363.1 | - | 3 - 4 | 330 | 106 | |
AY277595.1 | - | 3 - 4 | 358 | 106 | |
AY358674.1 | - | 3 - 4 | 358 | 106 | |
BC000642.2 | - | 3 - 4 | 369 | 106 | |
CR541735.1 | - | 2 - 3 | 279 | 106 | |
CR541760.1 | - | 2 - 3 | 279 | 106 | |
DA258969.1 | - | 3 - 4 | 387 | 106 |