The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101417.3 | NP_001094887.1 | 3 - 4 | 639 | 80 |
NM_001101426.3 | NP_001094896.1 | 4 - 5 | 789 | 80 | |
XM_006715770.3 | XP_006715833.1 | 4 - 5 | 959 | 80 | |
XM_011515499.2 | XP_011513801.1 | 4 - 5 | 934 | 80 | |
XM_011515501.2 | XP_011513803.1 | 4 - 5 | 962 | 80 | |
XM_011515508.2 | XP_011513810.1 | 4 - 5 | 1291 | 80 | |
XM_011515509.2 | XP_011513811.1 | 4 - 5 | 2140 | 80 | |
XM_017012575.1 | XP_016868064.1 | 4 - 5 | 934 | 80 | |
XM_017012576.1 | XP_016868065.1 | 4 - 5 | 946 | 80 | |
XM_017012577.1 | XP_016868066.1 | 3 - 4 | 1051 | 80 | |
XM_017012578.1 | XP_016868067.1 | 2 - 3 | 1466 | 80 | |
GenBank mRNA | DT932748.1 | - | 2 - 3 | 80 |