The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000906.3 | NP_000897.3 | 12 - 13 | 2353 | 65 |
XM_005245218.1 | XP_005245275.1 | 11 - 12 | 2346 | 65 | |
XM_017001374.1 | XP_016856863.1 | 12 - 13 | 2424 | 65 | |
GenBank mRNA | AK025024.1 | - | 12 - 13 | 2353 | 65 |
AK298090.1 | - | 4 - 5 | 376 | 65 | |
AK300446.1 | - | 11 - 12 | 1559 | 65 | |
BC063304.1 | - | 12 - 13 | 2402 | 65 | |
S72628.1 | - | 1 - 2 | 33 | 65 | |
X15357.1 | - | 12 - 13 | 1975 | 65 |