Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPOCK2
Entrez Gene ID
9806
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2
Gene Aliases
testican-2
Chromosome Location
Chr.10: 72059034 - 72089032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244950.1 NP_001231879.1 7 - 8 1153 79
NM_014767.2 NP_055582.1 8 - 9 1036 79
XM_011540404.2 XP_011538706.1 6 - 7 1367 79
GenBank mRNA AJ001453.1 - 7 - 8 994 79
AK292328.1 - 7 - 8 886 79
AY358921.1 - 7 - 8 906 79
BC023558.2 - 7 - 8 1147 79
D87465.1 - 8 - 9 1024 79

View Full Product Details