The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023011.3 | NP_075387.1 | 4 - 5 | 579 | 77 |
XM_006719991.3 | XP_006720054.1 | 3 - 4 | 454 | 77 | |
XM_011534845.2 | XP_011533147.1 | 4 - 5 | 1107 | 77 | |
XM_011534846.1 | XP_011533148.1 | 4 - 5 | 526 | 77 | |
XM_011534847.2 | XP_011533149.1 | 3 - 4 | 919 | 77 | |
XM_011534848.2 | XP_011533150.1 | 4 - 5 | 1064 | 77 | |
XM_017020709.1 | XP_016876198.1 | 3 - 4 | 1291 | 77 | |
XM_017020710.1 | XP_016876199.1 | 4 - 5 | 676 | 77 | |
GenBank mRNA | AF318575.1 | - | 4 - 5 | 472 | 77 |
AK092586.1 | - | 1 - 2 | 1327 | 77 | |
AK302003.1 | - | 4 - 5 | 515 | 77 | |
AY013250.1 | - | 4 - 5 | 579 | 77 |