Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16988143-16988546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4787841 c.2685G>T p.L895L Chr.1: 16988399-16988399 on Build GRCh38
6607274 c.2519G>A p.G840E Chr.1: 16988433-16988433 on Build GRCh38
899714 c.2789C>T p.S930L Chr.1: 16988208-16988208 on Build GRCh38
4484165 c.2777C>T p.S926F Chr.1: 16988220-16988220 on Build GRCh38
1295498 c.2630G>C p.G877A Chr.1: 16988454-16988454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985370872 Chr.1: 16988448-16988448 on Build GRCh38
rs796448884 Chr.1: 16988291-16988293 on Build GRCh38
rs755542233 Chr.1: 16988201-16988201 on Build GRCh38
rs576215702 Chr.1: 16988262-16988262 on Build GRCh38
rs922495404 Chr.1: 16988199-16988199 on Build GRCh38
rs367638044 Chr.1: 16988251-16988251 on Build GRCh38
rs375014566 Chr.1: 16988192-16988192 on Build GRCh38
rs763069105 Chr.1: 16988361-16988361 on Build GRCh38
rs756683120 Chr.1: 16988409-16988409 on Build GRCh38
rs775892776 Chr.1: 16988463-16988463 on Build GRCh38
rs368404440 Chr.1: 16988198-16988198 on Build GRCh38
rs190323680 Chr.1: 16988421-16988421 on Build GRCh38
rs779656439 Chr.1: 16988303-16988303 on Build GRCh38
rs886045578 Chr.1: 16988189-16988189 on Build GRCh38
rs146234171 Chr.1: 16988342-16988342 on Build GRCh38
rs757945614 Chr.1: 16988469-16988469 on Build GRCh38
rs749964479 Chr.1: 16988420-16988420 on Build GRCh38
rs755414129 Chr.1: 16988281-16988281 on Build GRCh38
rs751670298 Chr.1: 16988504-16988504 on Build GRCh38
rs114407349 Chr.1: 16988250-16988250 on Build GRCh38
rs745504274 Chr.1: 16988299-16988299 on Build GRCh38
rs199994961 Chr.1: 16988465-16988465 on Build GRCh38
rs376189607 Chr.1: 16988483-16988483 on Build GRCh38
rs12129596 Chr.1: 16988202-16988202 on Build GRCh38
rs767865035 Chr.1: 16988461-16988461 on Build GRCh38
rs760518104 Chr.1: 16988239-16988239 on Build GRCh38
rs766632717 Chr.1: 16988362-16988362 on Build GRCh38
rs567287489 Chr.1: 16988444-16988444 on Build GRCh38
rs200702121 Chr.1: 16988284-16988284 on Build GRCh38
rs138437271 Chr.1: 16988399-16988399 on Build GRCh38
rs567866687 Chr.1: 16988227-16988227 on Build GRCh38
rs746804111 Chr.1: 16988304-16988304 on Build GRCh38
rs370356637 Chr.1: 16988200-16988200 on Build GRCh38
rs371975975 Chr.1: 16988429-16988429 on Build GRCh38
rs200504984 Chr.1: 16988467-16988467 on Build GRCh38
rs755968778 Chr.1: 16988513-16988513 on Build GRCh38
rs778562581 Chr.1: 16988208-16988208 on Build GRCh38
rs144701072 Chr.1: 16988455-16988455 on Build GRCh38
rs777791931 Chr.1: 16988514-16988514 on Build GRCh38
rs749237785 Chr.1: 16988517-16988517 on Build GRCh38
rs765560804 Chr.1: 16988177-16988177 on Build GRCh38
rs924044483 Chr.1: 16988484-16988484 on Build GRCh38
rs564643512 Chr.1: 16988226-16988226 on Build GRCh38
rs769961390 Chr.1: 16988356-16988356 on Build GRCh38
rs79724242 Chr.1: 16988360-16988360 on Build GRCh38
rs9435662 Chr.1: 16988447-16988447 on Build GRCh38
rs749859324 Chr.1: 16988468-16988468 on Build GRCh38
rs752015774 Chr.1: 16988278-16988278 on Build GRCh38
rs372361807 Chr.1: 16988339-16988339 on Build GRCh38
rs771014262 Chr.1: 16988449-16988449 on Build GRCh38
rs561561604 Chr.1: 16988456-16988456 on Build GRCh38
rs765505590 Chr.1: 16988259-16988259 on Build GRCh38
rs371934965 Chr.1: 16988260-16988260 on Build GRCh38
rs764639673 Chr.1: 16988411-16988411 on Build GRCh38
rs145455452 Chr.1: 16988375-16988375 on Build GRCh38
rs767921922 Chr.1: 16988402-16988402 on Build GRCh38
rs538848816 Chr.1: 16988521-16988521 on Build GRCh38
rs754741241 Chr.1: 16988494-16988494 on Build GRCh38
rs368772418 Chr.1: 16988470-16988470 on Build GRCh38
rs775423066 Chr.1: 16988231-16988231 on Build GRCh38
rs746749073 Chr.1: 16988424-16988424 on Build GRCh38
rs762012587 Chr.1: 16988255-16988255 on Build GRCh38
rs768674400 Chr.1: 16988338-16988338 on Build GRCh38
rs370013359 Chr.1: 16988511-16988511 on Build GRCh38
rs758709320 Chr.1: 16988195-16988195 on Build GRCh38
rs769823261 Chr.1: 16988433-16988433 on Build GRCh38
rs748091717 Chr.1: 16988432-16988432 on Build GRCh38
rs34015442 Chr.1: 16988512-16988513 on Build GRCh38
rs530039964 Chr.1: 16988332-16988332 on Build GRCh38
rs560897844 Chr.1: 16988301-16988301 on Build GRCh38
rs141447403 Chr.1: 16988293-16988293 on Build GRCh38
rs376348581 Chr.1: 16988403-16988403 on Build GRCh38
rs181907428 Chr.1: 16988518-16988518 on Build GRCh38
rs754796671 Chr.1: 16988426-16988426 on Build GRCh38
rs143237568 Chr.1: 16988272-16988272 on Build GRCh38
rs148336170 Chr.1: 16988291-16988291 on Build GRCh38
rs556418739 Chr.1: 16988450-16988450 on Build GRCh38
rs3738815 Chr.1: 16988207-16988207 on Build GRCh38
rs763274429 Chr.1: 16988268-16988268 on Build GRCh38
rs781119190 Chr.1: 16988501-16988501 on Build GRCh38
rs201437622 Chr.1: 16988296-16988296 on Build GRCh38
rs755438239 Chr.1: 16988228-16988228 on Build GRCh38

View Full Product Details