Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF19
Entrez Gene ID
7567
Gene Name
zinc finger protein 19
Gene Aliases
KOX12
Chromosome Location
Chr.16: 71474073 - 71489351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK055567.1 - 2 - 3 91 82

View Full Product Details