Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCAPH2
Entrez Gene ID
29781
Gene Name
non-SMC condensin II complex subunit H2
Gene Aliases
CAPH2
Chromosome Location
Chr.22: 50508216 - 50524780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185011.1 NP_001171940.1 1 - 2 227 96
NM_014551.4 NP_055366.3 1 - 2 227 96
NM_152299.3 NP_689512.2 1 - 2 227 96
XM_011530685.2 XP_011528987.1 1 - 2 107 96
GenBank mRNA BC000473.2 - 1 - 2 200 96
BC001509.2 - 1 - 2 200 96
BC001833.2 - 1 - 2 200 96
BC001937.1 - 1 - 2 184 96
BC009441.1 - 1 - 2 183 96
CR456604.1 - 1 - 2 159 96
DB498781.1 - 1 - 2 228 96

View Full Product Details