Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FAS
- Entrez Gene ID
-
355
- Gene Name
- Fas cell surface death receptor
- Gene Aliases
-
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
- Chromosome Location
-
Chr.10: 88968429 - 89017061 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM922638.1 |
- |
|
401 |
|
Chr. - - [-] on Build |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
FAS antisense RNA 1 |
FAS-AS1 |
FAS-AS, FASAS, SAF |
-10.1kb
|
Anti-sense |
Chr. - 88991422 - 88992975 on Build GRCh38 |
Downstream |
microRNA 4679-2 |
MIR4679-2 |
|
52.7kb
|
Anti-sense |
Chr. - 89063335 - 89063411 on Build GRCh38 |
View Full Product Details