Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOCD
Entrez Gene ID
93649
Gene Name
myocardin
Gene Aliases
MYCD
Chromosome Location
Chr.17: 12665890 - 12768949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146312.2 NP_001139784.1 7 - 8 1015 63
NM_153604.3 NP_705832.1 7 - 8 1015 63
XM_005256863.1 XP_005256920.1 7 - 8 1015 63
XM_005256864.1 XP_005256921.1 8 - 9 1059 63
GenBank mRNA AF532596.1 - 7 - 8 716 63
AK097821.1 - 4 - 5 637 63
AK128608.1 - 7 - 8 915 63
AK292885.1 - 7 - 8 884 63
AY764180.1 - 7 - 8 1008 63
BC126307.1 - 7 - 8 774 63
BC143391.1 - 7 - 8 774 63

View Full Product Details