Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STRC
Entrez Gene ID
161497
Gene Name
stereocilin
Gene Aliases
DFNB16
Chromosome Location
Chr.15: 43599243 - 43619669 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153700.2 NP_714544.1 23 - 24 4621 89
XM_011521278.2 XP_011519580.1 24 - 25 5481 89
XM_017021954.1 XP_016877443.1 24 - 25 5478 89
XM_017021955.1 XP_016877444.1 24 - 25 5454 89
XM_017021956.1 XP_016877445.1 23 - 24 5418 89
XM_017021957.1 XP_016877446.1 24 - 25 5406 89
XM_017021958.1 XP_016877447.1 24 - 25 5403 89
XM_017021959.1 XP_016877448.1 24 - 25 5376 89
XM_017021962.1 XP_016877451.1 22 - 23 2678 89
GenBank mRNA BC129931.1 - 22 - 23 3467 89

View Full Product Details