The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002788.3 | NP_002779.1 | 1 - 2 | 130 | 68 |
NM_152132.2 | NP_687033.1 | 1 - 2 | 130 | 68 | |
GenBank mRNA | AK315158.1 | - | 1 - 2 | 79 | 68 |
BC005265.1 | - | 1 - 2 | 58 | 68 | |
BC029402.1 | - | 1 - 2 | 60 | 68 | |
BC038990.1 | - | 1 - 2 | 57 | 68 | |
CF128376.1 | - | 1 - 2 | 130 | 68 |