The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172070.3 | NP_742067.3 | 38 - 39 | 5551 | 92 |
XM_005246299.4 | XP_005246356.1 | 39 - 40 | 5641 | 92 | |
XM_005246300.4 | XP_005246357.1 | 39 - 40 | 5584 | 92 | |
XM_006712268.3 | XP_006712331.1 | 39 - 40 | 5638 | 92 | |
XM_006712269.3 | XP_006712332.1 | 38 - 39 | 5563 | 92 | |
XM_006712270.3 | XP_006712333.2 | 38 - 39 | 5497 | 92 | |
XM_006712271.3 | XP_006712334.1 | 38 - 39 | 5604 | 92 | |
GenBank mRNA | AB095944.1 | - | 9 - 10 | 1661 | 92 |
AK094741.1 | - | 2 - 3 | 323 | 92 | |
AK126998.1 | - | 16 - 17 | 2280 | 92 | |
AK127553.1 | - | 12 - 13 | 1557 | 92 | |
AK128490.1 | - | 22 - 23 | 3031 | 92 | |
AK303062.1 | - | 13 - 14 | 1651 | 92 | |
AL834144.1 | - | 10 - 11 | 1258 | 92 | |
BX649036.1 | - | 14 - 15 | 1787 | 92 | |
CR749469.1 | - | 16 - 17 | 2053 | 92 |