The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172193.2 | NP_751943.1 | 10 - 11 | 990 | 145 |
XM_011536422.2 | XP_011534724.1 | 10 - 11 | 913 | 145 | |
GenBank mRNA | AF111806.1 | - | 10 - 11 | 928 | 145 |
AK127202.1 | - | 10 - 11 | 913 | 145 | |
AL833437.1 | - | 5 - 6 | 420 | 145 | |
BC031270.1 | - | 10 - 11 | 953 | 145 | |
BC101595.1 | - | 10 - 11 | 990 | 145 | |
BC101597.1 | - | 10 - 11 | 990 | 145 |