The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173591.3 | NP_775862.3 | 50 - 51 | 6203 | 152 |
XM_005268802.3 | XP_005268859.1 | 54 - 55 | 7004 | 152 | |
XM_011538191.1 | XP_011536493.1 | 51 - 52 | 6280 | 152 | |
XM_011538192.2 | XP_011536494.1 | 48 - 49 | 6221 | 152 | |
XM_011538193.1 | XP_011536495.1 | 48 - 49 | 6140 | 152 | |
GenBank mRNA | AK096852.1 | - | 4 - 5 | 481 | 152 |
BC101017.2 | - | 4 - 5 | 352 | 152 | |
BC101018.1 | - | 4 - 5 | 352 | 152 |