Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUSC1-AS1
Entrez Gene ID
284618
Gene Name
RUSC1 antisense RNA 1
Gene Aliases
C1orf104
Chromosome Location
Chr.1: 155320460 - 155324147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK093295.1 - 1 - 2 414 104

View Full Product Details