The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281437.1 | NP_001268366.1 | 3 - 4 | 488 | 72 |
NM_001281438.1 | NP_001268367.1 | 3 - 4 | 492 | 72 | |
NM_007002.3 | NP_008933.2 | 4 - 5 | 609 | 72 | |
NM_175573.2 | NP_783163.1 | 4 - 5 | 605 | 72 | |
XM_005260257.1 | XP_005260314.1 | 5 - 6 | 696 | 72 | |
XM_011528503.1 | XP_011526805.1 | 4 - 5 | 1444 | 72 | |
XM_017027602.1 | XP_016883091.1 | 4 - 5 | 1444 | 72 | |
XM_017027603.1 | XP_016883092.1 | 5 - 6 | 696 | 72 | |
XM_017027604.1 | XP_016883093.1 | 4 - 5 | 1440 | 72 | |
XM_017027605.1 | XP_016883094.1 | 4 - 5 | 1444 | 72 | |
XM_017027606.1 | XP_016883095.1 | 3 - 4 | 1327 | 72 | |
XM_017027607.1 | XP_016883096.1 | 3 - 4 | 1323 | 72 | |
XM_017027608.1 | XP_016883097.1 | 2 - 3 | 1113 | 72 | |
XM_017027609.1 | XP_016883098.1 | 2 - 3 | 1113 | 72 | |
XM_017027610.1 | XP_016883099.1 | 2 - 3 | 1113 | 72 | |
GenBank mRNA | AK054985.1 | - | - | 1672 | 72 |
AK307459.1 | - | - | 539 | 72 | |
BC010733.1 | - | 1 - 2 | 121 | 72 | |
BC017245.2 | - | 4 - 5 | 505 | 72 | |
BG337785.1 | - | 3 - 4 | 448 | 72 | |
D64154.1 | - | 4 - 5 | 500 | 72 |