The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014555.3 | NP_055370.1 | 10 - 11 | 1631 | 87 |
XM_017017628.1 | XP_016873117.1 | 11 - 12 | 1718 | 87 | |
GenBank mRNA | AF177473.1 | - | 10 - 11 | 1631 | 87 |
AJ270996.1 | - | 10 - 11 | 1604 | 87 | |
BC093787.1 | - | 10 - 11 | 1628 | 87 | |
BC093789.1 | - | 10 - 11 | 1628 | 87 | |
BC143351.1 | - | 10 - 11 | 1628 | 87 | |
BC143353.1 | - | 10 - 11 | 1628 | 87 |